Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGAACATCAAGGGGTGAAACTGC[G/T]TGAAAACAGACATCAACAGCAAAGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MIER3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIER3 - MIER family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SETD9 - SET domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171990.2 | 699 | Missense Mutation | CGT,CTT | R,L 100 | NP_001165461.1 | |
NM_001323018.1 | 699 | Missense Mutation | CGT,CTT | R,L 74 | NP_001309947.1 | |
NM_001323019.1 | 699 | Missense Mutation | CGT,CTT | R,L 100 | NP_001309948.1 | |
NM_001323020.1 | 699 | Missense Mutation | CGT,CTT | R,L 74 | NP_001309949.1 | |
NM_001323022.1 | 699 | UTR 5 | NP_001309951.1 | |||
NM_153706.3 | 699 | Missense Mutation | CGT,CTT | R,L 100 | NP_714917.2 | |
XM_005248426.4 | 699 | Intron | XP_005248483.1 | |||
XM_011543139.2 | 699 | Missense Mutation | CGT,CTT | R,L 68 | XP_011541441.1 | |
XM_011543140.2 | 699 | Missense Mutation | CGT,CTT | R,L 24 | XP_011541442.1 |