Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCCTCACTGGTTTCCCCTTCCTC[C/T]GTGGGAGGTAGATGTTGAGGATTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EXD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXD1 - exonuclease 3'-5' domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286441.1 | 1187 | Silent Mutation | NP_001273370.1 | |||
NM_152596.3 | 1187 | Silent Mutation | NP_689809.2 | |||
XM_006720415.3 | 1187 | Silent Mutation | XP_006720478.1 | |||
XM_011521298.2 | 1187 | Silent Mutation | XP_011519600.1 | |||
XM_011521299.2 | 1187 | Silent Mutation | XP_011519601.1 | |||
XM_011521300.2 | 1187 | Silent Mutation | XP_011519602.1 | |||
XM_011521301.2 | 1187 | Silent Mutation | XP_011519603.1 | |||
XM_011521302.2 | 1187 | Silent Mutation | XP_011519604.1 | |||
XM_017021970.1 | 1187 | Silent Mutation | XP_016877459.1 | |||
XM_017021971.1 | 1187 | Silent Mutation | XP_016877460.1 |