Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATGTTATTCTTCTTTAATTTACCA[A/G]TGGCCATATCTTTCTGAAACACAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114240 MIM: 104180 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CAPN3 - calpain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000070.2 | Intron | NP_000061.1 | ||||
NM_024344.1 | Intron | NP_077320.1 | ||||
NM_173087.1 | Intron | NP_775110.1 | ||||
NM_173088.1 | Intron | NP_775111.1 | ||||
NM_173089.1 | Intron | NP_775112.1 | ||||
NM_173090.1 | Intron | NP_775113.1 |
GANC - glucosidase alpha, neutral C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |