Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTAGAGAGGTGTTTGGGGACTGTTA[A/G]CTCACTGGGCCTTTCTTCTATTCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608551 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
VPS18 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
VPS18 - VPS18, CORVET/HOPS core subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020857.2 | 651 | Intron | NP_065908.1 | |||
XM_011521843.2 | 651 | Intron | XP_011520145.1 | |||
XM_011521844.2 | 651 | UTR 5 | XP_011520146.1 | |||
XM_017022447.1 | 651 | Intron | XP_016877936.1 |
Set Membership: |
HapMap |