Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCATAGATGTTCCCAAGAATGTTG[C/T]TGGGGTAAGAAAAAAAGTGCTGTGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609610 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUBGCP4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TUBGCP4 - tubulin gamma complex associated protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286414.2 | Intron | NP_001273343.1 | ||||
NM_014444.4 | Intron | NP_055259.2 | ||||
XM_011521454.2 | Intron | XP_011519756.1 | ||||
XM_011521455.2 | Intron | XP_011519757.1 | ||||
XM_017022078.1 | Intron | XP_016877567.1 |
ZSCAN29 - zinc finger and SCAN domain containing 29 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |