Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCGGGGGCCTCCCGGGGAAGTCAT[C/T]CATCTGAATGTGGGAGGCAAGAGGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606214 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SHKBP1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SHKBP1 - SH3KBP1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138392.3 | 112 | Silent Mutation | ATC,ATT | I,I 21 | NP_612401.2 | |
XM_006723474.1 | 112 | Silent Mutation | ATC,ATT | I,I 21 | XP_006723537.1 | |
XM_006723475.1 | 112 | Silent Mutation | ATC,ATT | I,I 21 | XP_006723538.1 | |
XM_011527492.1 | 112 | Silent Mutation | ATC,ATT | I,I 21 | XP_011525794.1 | |
XM_011527493.1 | 112 | Intron | XP_011525795.1 | |||
XM_017027473.1 | 112 | Silent Mutation | ATC,ATT | I,I 21 | XP_016882962.1 | |
XM_017027474.1 | 112 | Intron | XP_016882963.1 |
SPTBN4 - spectrin beta, non-erythrocytic 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020971.2 | 112 | Intron | NP_066022.2 | |||
NM_025213.2 | 112 | Intron | NP_079489.2 | |||
XM_011527173.2 | 112 | Intron | XP_011525475.1 | |||
XM_017027049.1 | 112 | Intron | XP_016882538.1 | |||
XM_017027050.1 | 112 | Intron | XP_016882539.1 | |||
XM_017027051.1 | 112 | Intron | XP_016882540.1 | |||
XM_017027052.1 | 112 | Intron | XP_016882541.1 |
Set Membership: |
HapMap |