Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGTTCTCAGACTGCAGGTAGAC[A/C]AAAGTAGGTAGACAGAGGCCTTCTA
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||
Literature Links: |
ZNF285 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
ZNF285 - zinc finger protein 285 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291488.1 | Intron | NP_001278417.1 | ||||
NM_001291489.1 | Intron | NP_001278418.1 | ||||
NM_001291490.1 | Intron | NP_001278419.1 | ||||
NM_001291491.1 | Intron | NP_001278420.1 | ||||
NM_152354.5 | Intron | NP_689567.4 |