Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCGGGCCGCCGTCCGCGCCCCCG[A/G]GACTGGTCCGGGGCTGGCCCGCCCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611112 | ||||||||||||||||||||
Literature Links: |
DACT3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DACT3 - dishevelled binding antagonist of beta catenin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301046.1 | 1665 | Missense Mutation | CCC,CTC | P,L 47 | NP_001287975.1 | |
NM_145056.2 | 1665 | Missense Mutation | CCC,CTC | P,L 272 | NP_659493.2 | |
XM_011526499.2 | 1665 | Missense Mutation | CCC,CTC | P,L 47 | XP_011524801.1 | |
XM_011526500.2 | 1665 | Missense Mutation | CCC,CTC | P,L 47 | XP_011524802.1 | |
XM_017026334.1 | 1665 | Missense Mutation | CCC,CTC | P,L 204 | XP_016881823.1 |