Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCTCTGCCTCTCGTGGCCCCCT[A/C]GGTGCTGTATATGAAACTTGTTTAG
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 600530 | |||||||||||||||||||||||||||||
Literature Links: |
MFSD9 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
MFSD9 - major facilitator superfamily domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322080.1 | 2935 | UTR 3 | NP_001309009.1 | |||
NM_001322081.1 | 2935 | UTR 3 | NP_001309010.1 | |||
NM_032718.4 | 2935 | UTR 3 | NP_116107.3 | |||
XM_011512005.2 | 2935 | UTR 3 | XP_011510307.1 | |||
XM_011512006.2 | 2935 | UTR 3 | XP_011510308.1 | |||
XM_011512007.2 | 2935 | UTR 3 | XP_011510309.1 |
SLC9A2 - solute carrier family 9 member A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |