Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGAACCTGGTCTCCCTAGGTGA[A/G]GATAATGTCCGTCCAGAAGCCTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF534 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF534 - zinc finger protein 534 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143938.1 | 208 | Intron | NP_001137410.1 | |||
NM_001143939.1 | 208 | Silent Mutation | GAA,GAG | E,E 49 | NP_001137411.1 | |
NM_001291368.1 | 208 | Intron | NP_001278297.1 | |||
NM_001291369.1 | 208 | Intron | NP_001278298.1 |
Set Membership: |
HapMap |