Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGAACTTGGCTGACCTCCGCAG[C/G]TTCCGCCCGGTGAGGGACGTGAGTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612399 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TLE6 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TLE6 - transducin like enhancer of split 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143986.1 | 64 | UTR 5 | NP_001137458.1 | |||
NM_024760.2 | 64 | UTR 5 | NP_079036.1 | |||
XM_005259645.2 | 64 | UTR 5 | XP_005259702.1 | |||
XM_011528300.2 | 64 | Intron | XP_011526602.1 | |||
XM_011528301.1 | 64 | Intron | XP_011526603.1 | |||
XM_011528306.1 | 64 | Intron | XP_011526608.1 |
Set Membership: |
HapMap |