Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAAACGAAAAACGAAAGCTGAGT[G/T]CATAGAGTTGGAAAGGGGAGCGAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300160 | ||||||||||||||||||||
Literature Links: |
DDX3X PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DDX3X - DEAD-box helicase 3, X-linked | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193416.2 | 449 | Intron | NP_001180345.1 | |||
NM_001193417.2 | 449 | Intron | NP_001180346.1 | |||
NM_001356.4 | 449 | UTR 5 | NP_001347.3 | |||
XM_011543892.2 | 449 | Intron | XP_011542194.1 | |||
XM_017029313.1 | 449 | UTR 5 | XP_016884802.1 | |||
XM_017029314.1 | 449 | UTR 5 | XP_016884803.1 |
LOC107985678 - uncharacterized LOC107985678 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_017030007.1 | 449 | Intron | XP_016885496.1 |