Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACAATTGGTCAAACCACAAGAAC[A/G]CTGTTACCTTGAGCCTGAGAAGCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FGF13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FGF13 - fibroblast growth factor 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139498.1 | 1865 | UTR 3 | NP_001132970.1 | |||
NM_001139500.1 | 1865 | UTR 3 | NP_001132972.1 | |||
NM_001139501.1 | 1865 | UTR 3 | NP_001132973.1 | |||
NM_001139502.1 | 1865 | UTR 3 | NP_001132974.1 | |||
NM_004114.3 | 1865 | UTR 3 | NP_004105.1 | |||
NM_033642.2 | 1865 | UTR 3 | NP_378668.1 | |||
XM_005262399.1 | 1865 | UTR 3 | XP_005262456.1 |
Set Membership: |
HapMap |