Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAATTAGAGTCAGAATCATCAGA[A/C]GTGGATGGCTTCCTTTTGCGGCGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126430 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TOP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
TOP2A - topoisomerase (DNA) II alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001067.3 | 4511 | Silent Mutation | ACG,ACT | T,T 1470 | NP_001058.2 | |
XM_005257632.1 | 4511 | Silent Mutation | ACG,ACT | T,T 1458 | XP_005257689.1 | |
XM_011525165.1 | 4511 | Intron | XP_011523467.1 |
Set Membership: |
HapMap |