Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATGGCCCTGGTCAGCTATGCCAT[A/G]AACTTTGCCATCGGGTGTGTGGTCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602024 MIM: 610692 | ||||||||||||||||||||
Literature Links: |
CLCNKA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CLCNKA - chloride voltage-gated channel Ka | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042704.1 | 281 | Missense Mutation | ATA,ATG | I,M 67 | NP_001036169.1 | |
NM_001257139.1 | 281 | Missense Mutation | ATA,ATG | I,M 67 | NP_001244068.1 | |
NM_004070.3 | 281 | Missense Mutation | ATA,ATG | I,M 67 | NP_004061.3 |
HSPB7 - heat shock protein family B (small) member 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |