Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAATAATAAGAATAGCACTTTTC[A/T]TGAGTTCTACGAGTTGTTTCAGTTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300921 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMCX5-GPRASP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ARMCX5-GPRASP2 - ARMCX5-GPRASP2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199818.1 | Intron | NP_001186747.1 |
BHLHB9 - basic helix-loop-helix domain containing, class B, 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142524.1 | Intron | NP_001135996.1 | ||||
NM_001142525.1 | Intron | NP_001135997.1 | ||||
NM_001142526.1 | Intron | NP_001135998.1 | ||||
NM_001142527.1 | Intron | NP_001135999.1 | ||||
NM_001142528.1 | Intron | NP_001136000.1 | ||||
NM_001142529.1 | Intron | NP_001136001.1 | ||||
NM_001142530.1 | Intron | NP_001136002.1 | ||||
NM_030639.2 | Intron | NP_085142.1 |
GPRASP2 - G protein-coupled receptor associated sorting protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |