Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACACAGTGGCACTGACGACGGGAA[A/G]CCTGGGCTCGCTCCATCGATAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIGLEC16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SIGLEC16 - sialic acid binding Ig like lectin 16 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VRK3 - vaccinia related kinase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025778.1 | Intron | NP_001020949.1 | ||||
NM_001308420.1 | Intron | NP_001295349.1 | ||||
NM_016440.3 | Intron | NP_057524.3 | ||||
XM_005258971.3 | Intron | XP_005259028.1 | ||||
XM_005258972.3 | Intron | XP_005259029.1 | ||||
XM_006723237.3 | Intron | XP_006723300.1 | ||||
XM_006723238.3 | Intron | XP_006723301.1 | ||||
XM_011527023.2 | Intron | XP_011525325.1 |
Set Membership: |
HapMap |