Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACCCTAATCTAGGTGTTACATTTG[C/T]CTTTTTGTACTTCGTATTGAAATTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611541 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SNX27 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SNX27 - sorting nexin family member 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030918.5 | Intron | NP_112180.4 | ||||
XM_005245509.2 | Intron | XP_005245566.1 | ||||
XM_005245510.3 | Intron | XP_005245567.1 | ||||
XM_005245511.4 | Intron | XP_005245568.1 | ||||
XM_011510024.2 | Intron | XP_011508326.1 | ||||
XM_011510025.2 | Intron | XP_011508327.1 | ||||
XM_011510026.2 | Intron | XP_011508328.1 | ||||
XM_017002417.1 | Intron | XP_016857906.1 |
Set Membership: |
HapMap |