Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCACTGCACCTGGCCTAAAATGAA[C/T]GTCTTTATAGGTAACATCCAGACCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611984 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MRPS21 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MRPS21 - mitochondrial ribosomal protein S21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018997.3 | Intron | NP_061870.1 | ||||
NM_031901.5 | Intron | NP_114107.1 |
Set Membership: |
HapMap |