Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAATGTTGCCTACACAGCATTA[C/T]GAGTGTAATAATTTTCATTAGGTAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611656 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIKE1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SIKE1 - suppressor of IKBKE 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102396.1 | 1753 | UTR 3 | NP_001095866.1 | |||
NM_025073.2 | 1753 | UTR 3 | NP_079349.2 |