Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCGGAGTGGTGTCCAGAAACCA[A/G]CCACCTGAAAAATAGGTCTGTTGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SAMD4B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SAMD4B - sterile alpha motif domain containing 4B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303614.1 | Intron | NP_001290543.1 | ||||
NM_018028.3 | Intron | NP_060498.2 | ||||
XM_011527057.2 | Intron | XP_011525359.1 | ||||
XM_011527060.2 | Intron | XP_011525362.1 | ||||
XM_011527061.2 | Intron | XP_011525363.1 | ||||
XM_011527062.2 | Intron | XP_011525364.1 | ||||
XM_011527063.2 | Intron | XP_011525365.1 | ||||
XM_011527064.2 | Intron | XP_011525366.1 | ||||
XM_017026918.1 | Intron | XP_016882407.1 | ||||
XM_017026919.1 | Intron | XP_016882408.1 | ||||
XM_017026920.1 | Intron | XP_016882409.1 | ||||
XM_017026921.1 | Intron | XP_016882410.1 | ||||
XM_017026922.1 | Intron | XP_016882411.1 | ||||
XM_017026923.1 | Intron | XP_016882412.1 |