Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTGTGGAGTGATTTGCCCTGTCC[C/T]GTGGGCAGTGCTTGATTCCCAGCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600233 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GABRG3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GABRG3 - gamma-aminobutyric acid type A receptor gamma3 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270873.1 | Intron | NP_001257802.1 | ||||
NM_033223.4 | Intron | NP_150092.2 | ||||
XM_011521430.2 | Intron | XP_011519732.1 | ||||
XM_011521431.2 | Intron | XP_011519733.1 | ||||
XM_017022058.1 | Intron | XP_016877547.1 | ||||
XM_017022059.1 | Intron | XP_016877548.1 | ||||
XM_017022060.1 | Intron | XP_016877549.1 |