Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGCCCTTCACTTCTCTAGGCTAAT[C/T]TGATACGAATGTACTGTTCTTGAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611417 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SGSM1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SGSM1 - small G protein signaling modulator 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039948.3 | Intron | NP_001035037.1 | ||||
NM_001098497.2 | Intron | NP_001091967.1 | ||||
NM_001098498.2 | Intron | NP_001091968.1 | ||||
NM_133454.3 | Intron | NP_597711.1 |
Set Membership: |
HapMap |