Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAACAAAAGCATGTGAGGCTAGAC[A/G]CACACATTTTATCATCCTCAGTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108345 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NAT1 - N-acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000662.7 | Intron | NP_000653.3 | ||||
NM_001160170.3 | Intron | NP_001153642.1 | ||||
NM_001160171.3 | Intron | NP_001153643.1 | ||||
NM_001160172.3 | Intron | NP_001153644.1 | ||||
NM_001160173.3 | Intron | NP_001153645.1 | ||||
NM_001160174.2 | Intron | NP_001153646.1 | ||||
NM_001160175.3 | Intron | NP_001153647.1 | ||||
NM_001160176.3 | Intron | NP_001153648.1 | ||||
NM_001160179.2 | Intron | NP_001153651.1 | ||||
NM_001291962.1 | Intron | NP_001278891.1 | ||||
XM_006716410.3 | Intron | XP_006716473.1 | ||||
XM_011544687.1 | Intron | XP_011542989.1 | ||||
XM_011544688.1 | Intron | XP_011542990.1 | ||||
XM_011544689.2 | Intron | XP_011542991.1 | ||||
XM_017013947.1 | Intron | XP_016869436.1 |
Set Membership: |
HapMap |