Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAAGATGTGGCTACTTTTCTTGCA[C/T]CCATAAGGTTCTGCATCAGTATGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF790 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF790 - zinc finger protein 790 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242800.1 | 1850 | Silent Mutation | GGA,GGG | G,G 563 | NP_001229729.1 | |
NM_001242801.1 | 1850 | Silent Mutation | GGA,GGG | G,G 563 | NP_001229730.1 | |
NM_001242802.1 | 1850 | Silent Mutation | GGA,GGG | G,G 563 | NP_001229731.1 | |
NM_206894.3 | 1850 | Silent Mutation | GGA,GGG | G,G 563 | NP_996777.2 | |
XM_005258903.4 | 1850 | Silent Mutation | GGA,GGG | G,G 563 | XP_005258960.1 | |
XM_011526950.2 | 1850 | Silent Mutation | GGA,GGG | G,G 563 | XP_011525252.1 |
ZNF790-AS1 - ZNF790 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |