Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGTTCCTGCCGGCTGTATTCGGG[C/T]CTTGGACTGGACTGAGAAGCTACGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615470 MIM: 610884 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP89 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP89 - centrosomal protein 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032816.4 | Intron | NP_116205.3 | ||||
XM_005259344.3 | Intron | XP_005259401.1 | ||||
XM_011527425.2 | Intron | XP_011525727.1 | ||||
XM_017027398.1 | Intron | XP_016882887.1 |
FAAP24 - Fanconi anemia core complex associated protein 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300978.1 | Intron | NP_001287907.1 | ||||
NM_152266.4 | Intron | NP_689479.1 | ||||
XM_005259393.3 | Intron | XP_005259450.1 |
RHPN2 - rhophilin Rho GTPase binding protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |