Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCATACAAAAGCCATCAGTGATA[C/T]GATGTGGGTTCTTCATAAAAATATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611271 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF18A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF18A - kinesin family member 18A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
METTL15 - methyltransferase like 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001113528.1 | Intron | NP_001107000.1 | ||||
NM_001297775.1 | Intron | NP_001284704.1 | ||||
NM_152636.2 | Intron | NP_689849.2 | ||||
XM_011519935.2 | Intron | XP_011518237.1 | ||||
XM_011519937.2 | Intron | XP_011518239.1 | ||||
XM_011519940.2 | Intron | XP_011518242.1 | ||||
XM_011519941.2 | Intron | XP_011518243.1 | ||||
XM_017017297.1 | Intron | XP_016872786.1 | ||||
XM_017017298.1 | Intron | XP_016872787.1 | ||||
XM_017017299.1 | Intron | XP_016872788.1 | ||||
XM_017017300.1 | Intron | XP_016872789.1 | ||||
XM_017017301.1 | Intron | XP_016872790.1 | ||||
XM_017017302.1 | Intron | XP_016872791.1 | ||||
XM_017017303.1 | Intron | XP_016872792.1 |
Set Membership: |
HapMap |