Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGTGAGTTGTGCAATAAATGAGC[G/T]GTCACTCTGAACAGGTGGGAAAGAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603312 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BBOX1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BBOX1 - gamma-butyrobetaine hydroxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003986.2 | 237 | Intron | NP_003977.1 | |||
XM_005253159.2 | 237 | UTR 5 | XP_005253216.1 | |||
XM_005253160.2 | 237 | UTR 5 | XP_005253217.1 | |||
XM_005253161.2 | 237 | Intron | XP_005253218.1 | |||
XM_011520402.1 | 237 | UTR 5 | XP_011518704.1 | |||
XM_017018403.1 | 237 | Intron | XP_016873892.1 |
BBOX1-AS1 - BBOX1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |