Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCAAATGGGGTTTATCTAGAGAAT[A/C]CAAGACTGGCTTAAAGCCTGGTACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300482 | ||||||||||||||||||||
Literature Links: |
GAB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAB3 - GRB2 associated binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081573.2 | Intron | NP_001075042.1 | ||||
NM_001282283.1 | Intron | NP_001269212.1 | ||||
NM_080612.3 | Intron | NP_542179.1 | ||||
XM_005274648.1 | Intron | XP_005274705.1 | ||||
XM_006724804.1 | Intron | XP_006724867.1 | ||||
XM_011531103.1 | Intron | XP_011529405.1 | ||||
XM_011531105.1 | Intron | XP_011529407.1 | ||||
XM_011531106.1 | Intron | XP_011529408.1 | ||||
XM_017029276.1 | Intron | XP_016884765.1 |