Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTGTGCAGATTCAGCTTTTACAC[A/G]TTCCACTGTGGAGGAAGAAGAAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
59 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600413 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GBP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GBP3 - guanylate binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319179.1 | 1609 | Missense Mutation | CGT,TGT | R,C 412 | NP_001306108.1 | |
NM_001319180.1 | 1609 | Missense Mutation | CGT,TGT | R,C 357 | NP_001306109.1 | |
NM_001319181.1 | 1609 | Missense Mutation | CGT,TGT | R,C 464 | NP_001306110.1 | |
NM_018284.2 | 1609 | Missense Mutation | CGT,TGT | R,C 491 | NP_060754.2 | |
XM_005270743.2 | 1609 | Missense Mutation | CGT,TGT | R,C 412 | XP_005270800.1 | |
XM_006710574.2 | 1609 | Missense Mutation | CGT,TGT | R,C 464 | XP_006710637.1 | |
XM_011541233.1 | 1609 | Missense Mutation | CGT,TGT | R,C 412 | XP_011539535.1 | |
XM_011541234.1 | 1609 | Missense Mutation | CGT,TGT | R,C 410 | XP_011539536.1 | |
XM_017001013.1 | 1609 | Missense Mutation | CGT,TGT | R,C 357 | XP_016856502.1 | |
XM_017001014.1 | 1609 | Missense Mutation | CGT,TGT | R,C 276 | XP_016856503.1 |
Set Membership: |
HapMap |