Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAAACCAGCAGTGAACCCTGTAG[A/C]ATGCCTTGGATCCCAGCACTCAGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 126065 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP24A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CYP24A1 - cytochrome P450 family 24 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000782.4 | 2002 | UTR 3 | NP_000773.2 | |||
NM_001128915.1 | 2002 | UTR 3 | NP_001122387.1 | |||
XM_005260304.4 | 2002 | UTR 3 | XP_005260361.1 | |||
XM_017027691.1 | 2002 | UTR 3 | XP_016883180.1 | |||
XM_017027692.1 | 2002 | Intron | XP_016883181.1 | |||
XM_017027693.1 | 2002 | UTR 3 | XP_016883182.1 |