Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAATGTGCAAAACTGAATCTGTTT[A/C]TTTGGGGAGCACAAACCAGTAGAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602967 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927770 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101927770 - uncharacterized LOC101927770 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF217 - zinc finger protein 217 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006526.2 | Intron | NP_006517.1 | ||||
XM_005260545.3 | Intron | XP_005260602.1 | ||||
XM_006723875.2 | Intron | XP_006723938.1 | ||||
XM_011529036.1 | Intron | XP_011527338.1 | ||||
XM_017028059.1 | Intron | XP_016883548.1 |
Set Membership: |
HapMap |