Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATAATATTACAATGCTGTGATACA[G/T]ATTTATAAACACATATGCTGAAGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612784 MIM: 600215 MIM: 614550 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HYPK PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HYPK - huntingtin interacting protein K | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFAP1 - microfibrillar associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005926.2 | Intron | NP_005917.2 |
SERF2-C15ORF63 - SERF2-C15orf63 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERINC4 - serine incorporator 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |