Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCATCTAGAAGGGGTGCTCTGCAA[C/T]GTCTAGTAAATAACGGATTGCCTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612820 MIM: 615176 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPTN PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NPTN - neuroplastin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161363.1 | Intron | NP_001154835.1 | ||||
NM_001161364.1 | Intron | NP_001154836.1 | ||||
NM_012428.3 | Intron | NP_036560.1 | ||||
NM_017455.3 | Intron | NP_059429.1 | ||||
XM_011521452.2 | Intron | XP_011519754.1 |
NPTN-IT1 - NPTN intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REC114 - REC114 meiotic recombination protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |