Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TBC1D21 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TBC1D21 - TBC1 domain family member 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286434.1 | Intron | NP_001273363.1 | ||||
NM_153356.2 | Intron | NP_699187.1 | ||||
XM_006720409.3 | Intron | XP_006720472.1 | ||||
XM_006720410.3 | Intron | XP_006720473.1 | ||||
XM_006720411.3 | Intron | XP_006720474.1 | ||||
XM_011521281.2 | Intron | XP_011519583.1 | ||||
XM_011521282.2 | Intron | XP_011519584.1 | ||||
XM_011521283.2 | Intron | XP_011519585.1 | ||||
XM_011521284.2 | Intron | XP_011519586.1 |