Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAAAATTAGCGTATTATCTCAAT[C/T]AATATATGCAGCACACTAGGTCAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 120520 | |||||||||||||||||||||||
Literature Links: |
MME PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
MME - membrane metallo-endopeptidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000902.3 | Intron | NP_000893.2 | ||||
NM_007287.2 | Intron | NP_009218.2 | ||||
NM_007288.2 | Intron | NP_009219.2 | ||||
NM_007289.2 | Intron | NP_009220.2 | ||||
XM_006713646.3 | Intron | XP_006713709.1 | ||||
XM_006713647.3 | Intron | XP_006713710.1 | ||||
XM_011512855.2 | Intron | XP_011511157.1 | ||||
XM_011512856.2 | Intron | XP_011511158.1 | ||||
XM_011512857.2 | Intron | XP_011511159.1 |