Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAGGTCTTCAAAGCGTTAGATC[A/G]GTCAAGTCTTAGAGAATCAAAAAGT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606138 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CGRRF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CGRRF1 - cell growth regulator with ring finger domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006568.2 | Intron | NP_006559.1 | ||||
XM_011536362.2 | Intron | XP_011534664.1 | ||||
XM_017020931.1 | Intron | XP_016876420.1 | ||||
XM_017020932.1 | Intron | XP_016876421.1 |