Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCATAAATATCACACAGTGAAAAAT[C/T]TATCACAAACTAAATACAGTAACAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607974 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NETO2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NETO2 - neuropilin and tolloid like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001201477.1 | 3326 | UTR 3 | NP_001188406.1 | |||
NM_018092.4 | 3326 | UTR 3 | NP_060562.3 | |||
XM_006721289.1 | 3326 | UTR 3 | XP_006721352.1 | |||
XM_006721291.1 | 3326 | UTR 3 | XP_006721354.1 | |||
XM_006721292.1 | 3326 | Intron | XP_006721355.1 | |||
XM_017023740.1 | 3326 | UTR 3 | XP_016879229.1 |
Set Membership: |
HapMap |