Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCTGCGCTGGCCTCAAGGAACCA[C/G]GTTTAGACAAACTTCGGAGGCAAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120360 | ||||||||||||||||||||
Literature Links: |
MMP2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MMP2 - matrix metallopeptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127891.2 | Intron | NP_001121363.1 | ||||
NM_001302508.1 | Intron | NP_001289437.1 | ||||
NM_001302509.1 | Intron | NP_001289438.1 | ||||
NM_001302510.1 | Intron | NP_001289439.1 | ||||
NM_004530.5 | Intron | NP_004521.1 |