Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGTCACCAACTTCTAAACCTGG[C/T]ACTGGGCATTTGAAGAATAGGATTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603502 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IFRD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IFRD1 - interferon related developmental regulator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LSMEM1 - leucine rich single-pass membrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134468.1 | 687 | Intron | NP_001127940.1 | |||
NM_182597.2 | 687 | Intron | NP_872403.1 | |||
XM_011516074.1 | 687 | UTR 5 | XP_011514376.1 | |||
XM_011516075.2 | 687 | UTR 5 | XP_011514377.1 | |||
XM_011516076.2 | 687 | UTR 5 | XP_011514378.1 | |||
XM_017012028.1 | 687 | UTR 5 | XP_016867517.1 |