Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGCCCGGGCGGCGCGGGCGCGGGC[G/T]CTGCGGCGGGACAGCTCGACGCGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601430 | ||||||||||||||||||||
Literature Links: |
UPF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UPF1 - UPF1, RNA helicase and ATPase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297549.1 | 489 | Missense Mutation | GCT,TCT | A,S 69 | NP_001284478.1 | |
NM_002911.3 | 489 | Missense Mutation | GCT,TCT | A,S 69 | NP_002902.2 | |
XM_017027105.1 | 489 | Missense Mutation | GCT,TCT | A,S 69 | XP_016882594.1 | |
XM_017027106.1 | 489 | Missense Mutation | GCT,TCT | A,S 69 | XP_016882595.1 |