Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCCTGCTTCACAAGGAAGGATGA[A/G]TGGAGAATTGAGTGGAGTTACACGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607993 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUGP2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SUGP2 - SURP and G-patch domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017392.4 | 4310 | UTR 3 | NP_001017392.2 | |||
NM_001321697.1 | 4310 | Intron | NP_001308626.1 | |||
NM_001321698.1 | 4310 | Intron | NP_001308627.1 | |||
NM_001321699.1 | 4310 | UTR 3 | NP_001308628.1 | |||
NM_014884.4 | 4310 | Intron | NP_055699.2 | |||
XM_017026137.1 | 4310 | UTR 3 | XP_016881626.1 |
Set Membership: |
HapMap |