Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTGTCTCCTCTCTTCCATTCAA[A/G]TGGTACCCATGGATTACCAAGCTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194522 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF33B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ZNF33B - zinc finger protein 33B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305033.1 | Intron | NP_001291962.1 | ||||
NM_001305035.1 | Intron | NP_001291964.1 | ||||
NM_001305036.1 | Intron | NP_001291965.1 | ||||
NM_001305037.1 | Intron | NP_001291966.1 | ||||
NM_001305038.1 | Intron | NP_001291967.1 | ||||
NM_001305039.1 | Intron | NP_001291968.1 | ||||
NM_001305040.1 | Intron | NP_001291969.1 | ||||
NM_006955.2 | Intron | NP_008886.1 |
Set Membership: |
HapMap |