Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611448 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BMS1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
BMS1 - BMS1, ribosome biogenesis factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014753.3 | 112 | Intron | NP_055568.3 | |||
XM_005271846.3 | 112 | Intron | XP_005271903.1 | |||
XM_005271848.3 | 112 | Intron | XP_005271905.1 | |||
XM_005271849.3 | 112 | Intron | XP_005271906.1 | |||
XM_011540402.2 | 112 | Intron | XP_011538704.1 | |||
XM_011540403.2 | 112 | UTR 5 | XP_011538705.1 |