Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGTTATGGTATGTGAGGTATATG[A/G]TGAGTTTTGTTATGAGTCATAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151460 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRC PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTPRC - protein tyrosine phosphatase, receptor type C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267798.1 | Intron | NP_001254727.1 | ||||
NM_002838.4 | Intron | NP_002829.3 | ||||
NM_080921.3 | Intron | NP_563578.2 | ||||
XM_006711472.3 | Intron | XP_006711535.1 | ||||
XM_006711473.3 | Intron | XP_006711536.1 | ||||
XM_006711474.3 | Intron | XP_006711537.1 |
Set Membership: |
HapMap |