Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAACAGAAGAATTGATACTAGAC[C/T]TTCCCTCTCACTCATGGATTTGTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 136535 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMN1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
FMN1 - formin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001103184.3 | 9639 | UTR 3 | NP_001096654.1 | |||
NM_001277313.1 | 9639 | UTR 3 | NP_001264242.1 | |||
NM_001277314.1 | 9639 | Intron | NP_001264243.1 | |||
XM_011521504.2 | 9639 | UTR 3 | XP_011519806.1 | |||
XM_011521505.2 | 9639 | Intron | XP_011519807.1 | |||
XM_011521506.2 | 9639 | UTR 3 | XP_011519808.1 | |||
XM_011521507.2 | 9639 | Intron | XP_011519809.1 | |||
XM_011521509.2 | 9639 | UTR 3 | XP_011519811.1 | |||
XM_011521511.2 | 9639 | UTR 3 | XP_011519813.1 | |||
XM_017022130.1 | 9639 | UTR 3 | XP_016877619.1 | |||
XM_017022131.1 | 9639 | Intron | XP_016877620.1 | |||
XM_017022132.1 | 9639 | UTR 3 | XP_016877621.1 | |||
XM_017022133.1 | 9639 | UTR 3 | XP_016877622.1 | |||
XM_017022134.1 | 9639 | UTR 3 | XP_016877623.1 | |||
XM_017022135.1 | 9639 | UTR 3 | XP_016877624.1 |