Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTATATTTGAAGATCAAGGGCT[A/G]AAAATCCACTCAGTACCATTTAGCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116952 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDC42 - cell division cycle 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039802.1 | Intron | NP_001034891.1 | ||||
NM_001791.3 | Intron | NP_001782.1 | ||||
NM_044472.2 | Intron | NP_426359.1 |
Set Membership: |
HapMap |