Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGGGAGCCAGCGTGTGCTCCCCC[A/G]GGCGCACTGCAGGCCCCGGGAGGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605563 | ||||||||||||||||||||
Literature Links: |
CABP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CABP1 - calcium binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033677.1 | 486 | Missense Mutation | CAG,CGG | Q,R 140 | NP_001028849.1 | |
NM_004276.4 | 486 | Intron | NP_004267.2 | |||
NM_031205.3 | 486 | Intron | NP_112482.1 | |||
XM_017020235.1 | 486 | Missense Mutation | CAG,CGG | Q,R 140 | XP_016875724.1 | |
XM_017020236.1 | 486 | Intron | XP_016875725.1 | |||
XM_017020237.1 | 486 | Intron | XP_016875726.1 | |||
XM_017020238.1 | 486 | Intron | XP_016875727.1 | |||
XM_017020239.1 | 486 | Intron | XP_016875728.1 |