Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGGTTAAAGAGCACGGTCCCCTTG[T/A]GGTAGGCGTACTCCTTGGTACTGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300473 | ||||||||||||||||||||
Literature Links: |
NR0B1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NR0B1 - nuclear receptor subfamily 0 group B member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000475.4 | 1157 | Missense Mutation | CAC,CTC | H,L 381 | NP_000466.2 | |
XM_017029338.1 | 1157 | Missense Mutation | CAC,CTC | H,L 381 | XP_016884827.1 |