Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGTACCTAGACACTAGACCACAG[C/G]GACAGTTTTCTGCTGATTTTGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300120 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAMLD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MAMLD1 - mastermind like domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177465.2 | Intron | NP_001170936.1 | ||||
NM_001177466.2 | Intron | NP_001170937.1 | ||||
NM_005491.4 | Intron | NP_005482.2 | ||||
XM_006724802.3 | Intron | XP_006724865.1 | ||||
XM_006724803.3 | Intron | XP_006724866.1 | ||||
XM_011531092.2 | Intron | XP_011529394.1 | ||||
XM_011531093.2 | Intron | XP_011529395.1 | ||||
XM_011531095.2 | Intron | XP_011529397.1 | ||||
XM_017029188.1 | Intron | XP_016884677.1 | ||||
XM_017029189.1 | Intron | XP_016884678.1 |